MAT047024061 Matrix Scientific

4-Carbethoxyhexafluorobutyryl chloride

  • MDL number MFCD00054671
  • Cas Number : 18381-53-8
  • Molecular Weight : 286.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.47
Country of Origin USA
Boiling point 161-163
Product Link http://www.matrixscientific.com/005659.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/005659.html
Availability details for
Quantity
  • Availability
  • Add to Cart