MAT047024067 Matrix Scientific

3-(4-Methoxyphenyl)propionitrile

  • MDL number MFCD00016404
  • Cas Number : 22442-48-4
  • Molecular Weight : 161.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Density 1.05
Country of Origin USA
Boiling point 105�m
Product Link http://www.matrixscientific.com/005666.html

Safety Information

hcodes Irritant/Irritant-Harmful
MSDS http://www.matrixscientific.com/005666.html
Availability details for
Quantity
  • Availability
  • Add to Cart