MAT047024070 Matrix Scientific

3-(4-Hydroxyphenyl)propionitrile

  • MDL number MFCD00016492
  • Cas Number : 17362-17-3
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-59
Purity 98
Country of Origin US
Boiling point 166�Torr
Product Link https://www.matrixscientific.com/005669.html

Safety Information

hcodes Harmful-Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart