MAT047024076 Matrix Scientific

Dimethyl perfluoro-1,10-decanedicarboxylate

  • MDL number MFCD00236606
  • Cas Number : 84750-88-9
  • Molecular Weight : 618.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 33-40
Purity 96
Country of Origin USA
Product Link http://www.matrixscientific.com/005680.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005680.html
Availability details for
Quantity
  • Availability
  • Add to Cart