MAT047024077 Matrix Scientific

2'-Fluoro-4'-hydroxyacetophenone

  • MDL number MFCD00142713
  • Cas Number : 98619-07-9
  • Molecular Weight : 154.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-121
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/001479.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001479.html
Availability details for
Quantity
  • Availability
  • Add to Cart