MAT047024111 Matrix Scientific

4-Bromodiphenylmethane

  • MDL number MFCD00053165
  • Cas Number : 2116-36-1
  • Molecular Weight : 247.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-112
Density 1.34
Country of Origin USA
Boiling point 162
Product Link http://www.matrixscientific.com/005720.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005720.html
Availability details for
Quantity
  • Availability
  • Add to Cart