MAT047024124 Matrix Scientific

3'-Fluoropropiophenone

  • MDL number MFCD00061245
  • Cas Number : 455-67-4
  • Molecular Weight : 152.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 94-96�5 Torr
Product Link http://www.matrixscientific.com/005740.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005740.html
Availability details for
Quantity
  • Availability
  • Add to Cart