MAT047024145 Matrix Scientific

Methyl perfluorododecanoate

  • MDL number MFCD00236616
  • Cas Number : 56554-52-0
  • Molecular Weight : 628.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Density 1.66
Country of Origin USA
Product Link http://www.matrixscientific.com/005780.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005780.html
Availability details for
Quantity
  • Availability
  • Add to Cart