MAT047024188 Matrix Scientific

2,2,3,3,3-Pentafluoropropylaminehydrochloride

  • MDL number MFCD00054747
  • Cas Number : 374-14-1
  • Molecular Weight : 185.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 248
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/001548.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/001548.html
Availability details for
Quantity
  • Availability
  • Add to Cart