MAT047024196 Matrix Scientific

2,4-Bis(trifluoromethyl)benzoyl chloride

  • MDL number MFCD00061227
  • Cas Number : 53130-43-1
  • Molecular Weight : 276.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Boiling point 191-193
Product Link http://www.matrixscientific.com/005825.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/005825.html
Availability details for
Quantity
  • Availability
  • Add to Cart