MAT047024209 Matrix Scientific

2,4-Bis(trifluoromethyl)aniline

  • MDL number MFCD00236209
  • Cas Number : 367-71-5
  • Molecular Weight : 229.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 50�4 Torr
Product Link http://www.matrixscientific.com/005838.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/005838.html
Availability details for
Quantity
  • Availability
  • Add to Cart