MAT047024210 Matrix Scientific

2,4-Bis(trifluoromethyl)phenylacetic acid

  • MDL number MFCD00042492
  • Cas Number : 177952-39-5
  • Molecular Weight : 272.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-90
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/005840.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005840.html
Availability details for
Quantity
  • Availability
  • Add to Cart