MAT047024215 Matrix Scientific

2,6-Bis(trifluoromethyl)-4-hydroxyquinoline

  • MDL number MFCD00276581
  • Cas Number : 35877-04-4
  • Molecular Weight : 281.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 280-290
Country of Origin USA
Product Link http://www.matrixscientific.com/005846.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005846.html
Availability details for
Quantity
  • Availability
  • Add to Cart