MAT047024216 Matrix Scientific

5,7-Bis(trifluoromethyl)-2-(methylthio)pyrazolo-[1,5-a]pyrimidine-3-carboxamide

  • MDL number MFCD00174394
  • Cas Number : 175203-36-8
  • Molecular Weight : 344.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/005848.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005848.html
Availability details for
Quantity
  • Availability
  • Add to Cart