MAT047024224 Matrix Scientific

3,5-Bis(trifluoromethyl)benzhydrazide

  • MDL number MFCD00051848
  • Cas Number : 26107-82-4
  • Molecular Weight : 272.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-134
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/005859.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005859.html
Availability details for
Quantity
  • Availability
  • Add to Cart