MAT047024246 Matrix Scientific

3,5-Bis(trifluoromethyl)-N-ethylaniline, tech.

  • MDL number MFCD00203993
  • Cas Number : 49850-16-0
  • Molecular Weight : 257.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 96-98�mm
Product Link http://www.matrixscientific.com/005883.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005883.html
Availability details for
Quantity
  • Availability
  • Add to Cart