MAT047024273 Matrix Scientific

2,6-Dinitro-4-trifluoromethylbenzenesulfonic acidsodium salt

  • MDL number MFCD00059068
  • Cas Number : 54495-25-9
  • Molecular Weight : 338.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/005916.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005916.html
Availability details for
Quantity
  • Availability
  • Add to Cart