MAT047024296 Matrix Scientific

Tetrafluorobenzene-1,3-diol

  • MDL number MFCD00040202
  • Cas Number : 16840-25-8
  • Molecular Weight : 182.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-96
Purity 97
Country of Origin USA
Boiling point 217-218
Product Link http://www.matrixscientific.com/005943.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005943.html
Availability details for
Quantity
  • Availability
  • Add to Cart