MAT047024314 Matrix Scientific

4-(4-Fluorobenzyloxy)benzaldehyde

  • MDL number MFCD00173650
  • Cas Number : 56442-17-2
  • Molecular Weight : 230.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99-100
Country of Origin USA
Product Link http://www.matrixscientific.com/005961.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/005961.html
Availability details for
Quantity
  • Availability
  • Add to Cart