MAT047024345 Matrix Scientific

3-(Difluoromethoxy)benzonitrile

  • MDL number MFCD00236225
  • Cas Number : 97582-88-2
  • Molecular Weight : 169.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 85-95�4 Torr
Product Link http://www.matrixscientific.com/006001.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/006001.html
Availability details for
Quantity
  • Availability
  • Add to Cart