MAT047024348 Matrix Scientific

3-(Difluoromethoxy)benzylamine

  • MDL number MFCD00236230
  • Cas Number : 244022-71-7
  • Molecular Weight : 173.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient/argon
Country of Origin USA
Product Link http://www.matrixscientific.com/006006.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006006.html
Availability details for
Quantity
  • Availability
  • Add to Cart