MAT047024356 Matrix Scientific

4-(Difluoromethoxy)phenylacetonitrile

  • MDL number MFCD00236242
  • Cas Number : 41429-16-7
  • Molecular Weight : 183.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/006013.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/006013.html
Availability details for
Quantity
  • Availability
  • Add to Cart