MAT047024359 Matrix Scientific

4-(Difluoromethoxy)toluene

  • MDL number MFCD00042253
  • Cas Number : 1583-83-1
  • Molecular Weight : 158.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 165-167
Product Link http://www.matrixscientific.com/006016.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006016.html
Availability details for
Quantity
  • Availability
  • Add to Cart