MAT047024373 Matrix Scientific

3-Trifluoromethylbutyric acid

  • MDL number MFCD00039533
  • Cas Number : 348-75-4
  • Molecular Weight : 156.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Boiling point 90-91�mm
Product Link http://www.matrixscientific.com/006029.html

Safety Information

hcodes Corrosive/Stench
MSDS http://www.matrixscientific.com/006029.html
Availability details for
Quantity
  • Availability
  • Add to Cart