MAT047024383 Matrix Scientific

2'-Chloro-6'-fluoroacetophenone

  • MDL number MFCD01631390
  • Cas Number : 87327-69-3
  • Molecular Weight : 172.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/001511.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001511.html
Availability details for
Quantity
  • Availability
  • Add to Cart