MAT047024394 Matrix Scientific

4'-Iodo-2'-(trifluoromethyl)acetanilide

  • MDL number MFCD00052845
  • Cas Number : 97760-98-0
  • Molecular Weight : 329.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-136
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/001065.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001065.html
Availability details for
Quantity
  • Availability
  • Add to Cart