MAT047024398 Matrix Scientific

3-(2-Fluorophenoxy)benzyl bromide

  • MDL number MFCD00059914
  • Cas Number : 242812-04-0
  • Molecular Weight : 281.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.46
Country of Origin USA
Product Link http://www.matrixscientific.com/006063.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006063.html
Availability details for
Quantity
  • Availability
  • Add to Cart