MAT047024405 Matrix Scientific

5-(4-Chlorophenyl)-2-(trifluoromethyl)furan-3-carbonyl chloride

  • MDL number MFCD00275554
  • Cas Number : 175276-61-6
  • Molecular Weight : 309.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-137
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/006070.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006070.html
Availability details for
Quantity
  • Availability
  • Add to Cart