MAT047024413 Matrix Scientific

2-(4-Chlorophenyl)-3-(trifluoromethyl)pyrazole-4-carbonyl chloride

  • MDL number MFCD00068143
  • Cas Number : 175137-19-6
  • Molecular Weight : 309.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-53
Country of Origin USA
Boiling point 120�3mm
Product Link http://www.matrixscientific.com/006083.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/006083.html
Availability details for
Quantity
  • Availability
  • Add to Cart