MAT047024417 Matrix Scientific

4-Oxo-6-(trifluoromethyl)-1,4-dihydro-3-quinolinecarboxylic acid

  • MDL number MFCD00206334
  • Cas Number : 49713-47-5
  • Molecular Weight : 257.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/001514.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001514.html
Availability details for
Quantity
  • Availability
  • Add to Cart