MAT047024428 Matrix Scientific

4-Hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylic acid

  • MDL number MFCD00219848
  • Cas Number : 175203-86-8
  • Molecular Weight : 273.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/001515.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001515.html
Availability details for
Quantity
  • Availability
  • Add to Cart