MAT047024430 Matrix Scientific

1-(3-Fluorophenyl)imidazole

  • MDL number MFCD00060486
  • Cas Number : 25372-42-3
  • Molecular Weight : 162.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Boiling point 80�1mm
Product Link http://www.matrixscientific.com/006100.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006100.html
Availability details for
Quantity
  • Availability
  • Add to Cart