MAT047024439 Matrix Scientific

4-Hydroxy-6-(trifluoromethoxy)-2-(trifluoromethyl)quinoline

  • MDL number MFCD00276584
  • Cas Number : 125647-79-2
  • Molecular Weight : 297.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229-232
Country of Origin USA
Product Link http://www.matrixscientific.com/001517.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001517.html
Availability details for
Quantity
  • Availability
  • Add to Cart