MAT047024464 Matrix Scientific

Heptafluoropropyl 1,2,2,2-tetrafluoroethyl ether

  • MDL number MFCD00042308
  • Cas Number : 3330-15-2
  • Molecular Weight : 286.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.538
Storage temp. Refrigerate
Country of Origin USA
Boiling point 40-42
Product Link http://www.matrixscientific.com/006134.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/006134.html
Availability details for
Quantity
  • Availability
  • Add to Cart