MAT047024468 Matrix Scientific

4-Chloropropylbenzene

  • MDL number MFCD00270099
  • Cas Number : 52944-34-0
  • Molecular Weight : 154.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.0340/20
Country of Origin USA
Boiling point 190-192
Product Link http://www.matrixscientific.com/006150.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006150.html
Availability details for
Quantity
  • Availability
  • Add to Cart