MAT047024501 Matrix Scientific

Perfluorosebaconitrile

  • MDL number MFCD00042366
  • Cas Number : 2342-09-8
  • Molecular Weight : 452.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.673/20
Country of Origin USA
Boiling point 147-148
Product Link http://www.matrixscientific.com/006191.html

Safety Information

hcodes Irritant-Harmful
MSDS http://www.matrixscientific.com/006191.html
Availability details for
Quantity
  • Availability
  • Add to Cart