MAT047024505 Matrix Scientific

Isopropyl 4,4,4-trifluoroacetoacetate

  • MDL number MFCD00040990
  • Cas Number : 175230-50-9
  • Molecular Weight : 198.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Boiling point 42�mm
Product Link http://www.matrixscientific.com/001013.html

Safety Information

hcodes Flammable
MSDS http://www.matrixscientific.com/001013.html
Availability details for
Quantity
  • Availability
  • Add to Cart