MAT047024538 Matrix Scientific

1H,1H-Pentadecafluorooctyl acrylate

  • MDL number MFCD00039248
  • Cas Number : 307-98-2
  • Molecular Weight : 454.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 1.403
Flash Point 91
Country of Origin USA
Boiling point 188�0 Torr
Product Link http://www.matrixscientific.com/006251.html

Safety Information

hcodes Irritant/Refrigerate/Lachrymator
MSDS http://www.matrixscientific.com/006251.html
Availability details for
Quantity
  • Availability
  • Add to Cart