MAT047024547 Matrix Scientific

Ethyl pentafluorobenzoate

  • MDL number MFCD00039211
  • Cas Number : 4522-93-4
  • Molecular Weight : 240.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.431
Country of Origin USA
Boiling point 113
Product Link http://www.matrixscientific.com/006260.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006260.html
Availability details for
Quantity
  • Availability
  • Add to Cart