MAT047024579 Matrix Scientific

1-n-Hexyltheobromine

  • MDL number MFCD00041424
  • Cas Number : 1028-33-7
  • Molecular Weight : 264.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82-83
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/006295.html

Safety Information

hcodes Irritant-Harmful
MSDS http://www.matrixscientific.com/006295.html
Availability details for
Quantity
  • Availability
  • Add to Cart