MAT047024589 Matrix Scientific

2-(Methylthio)nicotinic acid hydrazide

  • MDL number MFCD01310832
  • Cas Number : 232926-33-9
  • Molecular Weight : 183.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-144
Country of Origin USA
Product Link http://www.matrixscientific.com/006310.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006310.html
Availability details for
Quantity
  • Availability
  • Add to Cart