MAT047024610 Matrix Scientific

1H,1H,11H-Eicosafluoro-1-undecanol, tech.

  • MDL number MFCD00039628
  • Cas Number : 307-70-0
  • Molecular Weight : 532.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-97
Purity 85
Country of Origin USA
Boiling point 180-181�0mm
Product Link http://www.matrixscientific.com/001536.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001536.html
Availability details for
Quantity
  • Availability
  • Add to Cart