MAT047024636 Matrix Scientific

4,4'-Bis(trifluoromethyl)-2,2'-dinitrodiphenyldisulfide

  • MDL number MFCD00018043
  • Cas Number : 860-39-9
  • Molecular Weight : 444.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-161
Purity 97
Country of Origin US
Product Link http://www.matrixscientific.com/006374.html

Safety Information

MSDS http://www.matrixscientific.com/006374.html
Availability details for
Quantity
  • Availability
  • Add to Cart