MAT047024640 Matrix Scientific

2,2'-Bis(trifluoromethyl)benzidine

  • MDL number MFCD00190155
  • Cas Number : 341-58-2
  • Molecular Weight : 320.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/006378.html

Safety Information

hcodes Irritant-Harmful
MSDS http://www.matrixscientific.com/006378.html
Availability details for
Quantity
  • Availability
  • Add to Cart