MAT047024647 Matrix Scientific

Ethyl 3-(trifluoromethyl)benzoate

  • MDL number MFCD00013560
  • Cas Number : 76783-59-0
  • Molecular Weight : 218.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 100-101� Torr
Product Link http://www.matrixscientific.com/006386.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006386.html
Availability details for
Quantity
  • Availability
  • Add to Cart