MAT047024649 Matrix Scientific

Ethyl 4-(trifluoromethyl)benzoate

  • MDL number MFCD00013563
  • Cas Number : 583-02-8
  • Molecular Weight : 218.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 81�Torr
Product Link http://www.matrixscientific.com/006388.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006388.html
Availability details for
Quantity
  • Availability
  • Add to Cart