MAT047024666 Matrix Scientific

2,7-Diacetylfluorene

  • MDL number MFCD00045332
  • Cas Number : 961-27-3
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182
Purity 98
Density 1.183
Flash Point 171
Country of Origin USA
Boiling point 459
Product Link http://www.matrixscientific.com/006405.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006405.html
Availability details for
Quantity
  • Availability
  • Add to Cart