MAT047024692 Matrix Scientific

Nitrosonium hexafluorophosphate

  • MDL number MFCD00040324
  • Cas Number : 16921-91-8
  • Molecular Weight : 174.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Refrigerate/argon
Country of Origin USA
Product Link http://www.matrixscientific.com/006437.html

Safety Information

hcodes Corrosive/Stored under Argon/Hygroscopic/Refrigerate
MSDS http://www.matrixscientific.com/006437.html
Availability details for
Quantity
  • Availability
  • Add to Cart