MAT047024734 Matrix Scientific

3'-(Trifluoromethoxy)acetophenone

  • MDL number MFCD00042403
  • Cas Number : 170141-63-6
  • Molecular Weight : 204.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.285
Country of Origin USA
Boiling point 49-51�mm
Product Link http://www.matrixscientific.com/006486.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/006486.html
Availability details for
Quantity
  • Availability
  • Add to Cart