MAT047024746 Matrix Scientific

5H-Octafluoropentanoic acid

  • MDL number MFCD00054690
  • Cas Number : 376-72-7
  • Molecular Weight : 246.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 0
Purity 97
Density 1.725
Country of Origin USA
Boiling point 155-160
Product Link http://www.matrixscientific.com/006498.html

Safety Information

hcodes Corrosive
MSDS http://www.matrixscientific.com/006498.html
Availability details for
Quantity
  • Availability
  • Add to Cart